Tisk…

α-Bromo-2-chlorophenylacetic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR11047-100G OR11047-25G OR11047-500G
APOSOR11047-100GEA 1660 CZK
APOSOR11047-100G APOSOR11047-25G APOSOR11047-500G
α-Bromo-2-chlorophenylacetic acid
α-Bromo-2-chlorophenylacetic acid
Vzorec: C₈H₆BrClO₂
Bod tání: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD03428513
CAS číslo: 29270-30-2
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR