Tisk…

2-Amino-5-chloronicotinohydrazide

Dodavatel: Apollo Scientific
Varování

OR110366-250MG
APOSOR110366-250MGEA 1660 CZK
APOSOR110366-250MG
2-Amino-5-chloronicotinohydrazide
2-Amino-5-chloronicotinohydrazide
Vzorec: C₆H₇ClN₄O
Storage Temperature: Ambient
MDL Number: MFCD16631261
CAS číslo: 1314959-58-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR