Tisk…

4-(5-Propyl-1,3,4-oxadiazol-2-yl)aniline

Dodavatel: Apollo Scientific

OR110246-1G OR110246-250MG
APOSOR110246-1GEA 7280 CZK
APOSOR110246-1G APOSOR110246-250MG
4-(5-Propyl-1,3,4-oxadiazol-2-yl)aniline
4-(5-Propyl-1,3,4-oxadiazol-2-yl)aniline
Vzorec: C₁₁H₁₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD12406816
CAS číslo: 1227954-86-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR