Tisk…

Ethyl (2-methyl-1,3-thiazol-4-yl)acetate

Dodavatel: Apollo Scientific

OR110217-5G OR110217-25G
APOSOR110217-5GEA 3540 CZK
APOSOR110217-5G APOSOR110217-25G
Ethyl (2-methyl-1,3-thiazol-4-yl)acetate
Ethyl (2-methyl-1,3-thiazol-4-yl)acetate
Vzorec: C₈H₁₁NO₂S
Bod varu: 114 °C
Hustota: 1,174 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00052519
CAS číslo: 37128-24-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR