Tisk…

2-(Methylthio)phenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid

OR10566-100G OR10566-25G
APOSOR10566-100GEA 8570 CZK
APOSOR10566-100G APOSOR10566-25G
2-(Methylthio)phenylboronic acid 97%
2-(Methylthio)phenylboronic acid
Vzorec: C₇H₉BO₂S
MW: 168.02
Bod tání: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD01318165
CAS číslo: 168618-42-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR