Tisk…

4-Hydroxyphenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

OR10505-25G OR10505-5G OR10505-100G
APOSOR10505-25GEA 992 CZK
APOSOR10505-25G APOSOR10505-5G APOSOR10505-100G
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Vzorec: C₆H₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS číslo: 71597-85-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR