Tisk…

4-(Benzyloxycarbonyl)benzeneboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-((Benzyloxy)carbonyl)phenyl)boronic acid, 4-Benzyloxycarbonylphenylboronic acid, 4-Carbobenzoxybenzeneboronic acid, 4-(Benzyloxycarbonyl)benzeneboronic acid, 4-Carbobenzoxyphenylboronic acid, 4-(Benzyloxycarbonyl)phenylboronic acid

OR10359-1G OR10359-25G OR10359-5G
APOSOR10359-1GEA 1080 CZK
APOSOR10359-1G APOSOR10359-25G APOSOR10359-5G
4-(Benzyloxycarbonyl)benzeneboronic acid 97%
4-(Benzyloxycarbonyl)benzeneboronic acid
Vzorec: C₁₄H₁₃BO₄
MW: 256.06 g/mol
Bod tání: 187…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179442
CAS číslo: 184000-11-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR