Tisk…

(4-Boc-aminophenyl)boronic acid 98%

Dodavatel: Apollo Scientific
Varování

OR10347-1G OR10347-25G OR10347-5G
APOSOR10347-1GEA 622 CZK
APOSOR10347-1G APOSOR10347-25G APOSOR10347-5G
(4-Boc-aminophenyl)boronic acid 98%
(4-Boc-aminophenyl)boronic acid
Vzorec: C₁₁H₁₆BNO₄
Storage Temperature: Ambient
MDL Number: MFCD02093054
CAS číslo: 380430-49-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR