Tisk…

4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride

Dodavatel: Apollo Scientific
Varování

OR10332-1G OR10332-250MG
APOSOR10332-1GEA 4960 CZK
APOSOR10332-1G APOSOR10332-250MG
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Vzorec: C₁₃H₂₀BNO₂·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD02179455
CAS číslo: 850568-55-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR