Tisk…

L(+)-Diethyl tartrate

Dodavatel: Apollo Scientific
Varování

Synonyms: (+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate

OR10049-100G OR10049-500G
APOSOR10049-100GEA 870 CZK
APOSOR10049-100G APOSOR10049-500G
L(+)-Diethyl tartrate
L(+)-Diethyl tartrate

Asymmetric synthesis catalyst; Chiral intermediate for pharmaceuticals and agrochemicals.

Vzorec: C₈H₁₄O₆
MW: 206.2
Bod varu: 280 °C
Bod tání: 17 °C
Hustota: 1.204 g/cm³ (25 °C)
Bod vzplanutí: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009143
CAS číslo: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR