Tisk…

N-Methylindole-6-carboxylic acid methyl ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 1-methyl-1H-indole-6-carboxylate, Methyl 1-Methylindole-6-carboxylate

OR0195-1G OR0195-5G OR0195-10G
APOSOR0195-1GEA 622 CZK
APOSOR0195-1G APOSOR0195-5G APOSOR0195-10G
N-Methylindole-6-carboxylic acid methyl ester 98%
N-Methylindole-6-carboxylic acid methyl ester
Vzorec: C₁₁H₁₁NO₂
MW: 189.22 g/mol
Bod tání: 90…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06245515
CAS číslo: 1204-32-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR