Tisk…

2-Carbomethoxybenzylamine hydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-(aminomethyl)benzoate hydrochloride

OR0170-1G OR0170-5G OR0170-10G
APOSOR0170-1GEA 1240 CZK
APOSOR0170-1G APOSOR0170-5G APOSOR0170-10G
2-Carbomethoxybenzylamine hydrochloride 95+%
2-Carbomethoxybenzylamine hydrochloride
Vzorec: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Bod tání: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD06245496
CAS číslo: 849020-92-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR