Tisk…

N-Cbz-L-asparagine 97%

Dodavatel: Apollo Scientific

OR01678-100G OR01678-25G OR01678-500G
APOSOR01678-100GEA 1820 CZK
APOSOR01678-100G APOSOR01678-25G APOSOR01678-500G
N-Cbz-L-asparagine 97%
N-Cbz-L-asparagine
Vzorec: C₁₂H₁₄N₂O₅
Bod tání: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00008035
CAS číslo: 2304-96-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR