Tisk…

5,8-Dihydroxy-1,4-naphthoquinone tech

Dodavatel: Apollo Scientific
Varování

Synonyms: Naphthazarin, 5,8-Dihydroxynaphthalene-1,4-dione

OR01425-1G
APOSOR01425-1GEA 6020 CZK
APOSOR01425-1G
5,8-Dihydroxy-1,4-naphthoquinone tech
5,8-Dihydroxy-1,4-naphthoquinone
Vzorec: C₁₀H₆O₄
MW: 190.16 g/mol
MDL Number: MFCD00001685
CAS číslo: 475-38-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR