Tisk…

Copper(I) thiocyanate

Dodavatel: Apollo Scientific
Varování

IN9847-250G IN9847-50G
APOSIN9847-250GEA 4900 CZK
APOSIN9847-250G APOSIN9847-50G
Copper(I) thiocyanate
Copper(I) thiocyanate
Vzorec: CCuNS
Bod tání: 1084 °C
Hustota: 2,84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010980
CAS číslo: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9+EHS,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR