Tisk…

Vanadium(II) chloride 85%

Dodavatel: Apollo Scientific
Nebezpečí

IN3760-5G
APOSIN3760-5GEA 13230 CZK
APOSIN3760-5G
Vanadium(II) chloride 85%
Vanadium(II) chloride
Vzorec: Cl₂V
MW: 121.85
Bod varu: 1506
Bod tání: 910 °C (subl.)
Hustota: 3.09 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS číslo: 10580-52-6
EINECS: 234-176-7
UN: 3260
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR