Tisk…

Palladium hydroxide on carbon 95% 20% Pd, wet support, powder

Dodavatel: Apollo Scientific
Varování

IN2807-5G IN2807-25G
APOSIN2807-5GEA 2610 CZK
APOSIN2807-5G APOSIN2807-25G
Palladium hydroxide on carbon 95% 20% Pd, wet support, powder
Palladium hydroxide
Vzorec: H₂O₂Pd
MW: 140.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS číslo: 12135-22-7
EINECS: 235-219-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR