Tisk…

Chlorid paladnatý 98%

Dodavatel: Apollo Scientific
Nebezpečí

IN2804-250MG
APOSIN2804-250MGEA 1240 CZK
APOSIN2804-250MG
Chlorid paladnatý 98%
Chlorid paladnatý

  • Less volume needed than for ethanol precipitation
  • Useful for precipitations with large sample volumes

Vzorec: PdCl₂
Bod tání: 678…680 °C
Hustota: 4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS číslo: 7647-10-1
EINECS: 231-596-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR