Tisk…

Iron(II) acetate, anhydrous

Dodavatel: Apollo Scientific
Varování

IN2041-25G
APOSIN2041-25GEA 6220 CZK
APOSIN2041-25G
Iron(II) acetate, anhydrous
Iron(II) acetate
Vzorec: C₄H₆FeO₄
MW: 173.94
Bod tání: 190…200 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00058909
CAS číslo: 3094-87-9
EINECS: 221-441-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR