Tisk…

Erbium tribromide nonahydrate 99.99%

Dodavatel: Apollo Scientific

Synonyms: Erbium (III) bromide nonahydrate

IN1687-100G IN1687-25G
APOSIN1687-100GEA 10550 CZK
APOSIN1687-100G APOSIN1687-25G
Erbium tribromide nonahydrate 99.99%
Erbium tribromide nonahydrate
Vzorec: Br₃Er·9H₂O
Bod tání: 950 °C
Storage Temperature: Ambient
MDL Number: MFCD00209649
CAS číslo: 29843-93-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR