Tisk…

Kyanokobalamin USP

Dodavatel: Apollo Scientific
Varování

BIV6236-1G BIV6236-250MG BIV6236-5G
APOSBIV6236-1GEA 1870 CZK
APOSBIV6236-1G APOSBIV6236-250MG APOSBIV6236-5G
Kyanokobalamin USP
Kyanokobalamin
Vzorec: C₆₃H₈₈CoN₁₄O₁₄P
Bod tání: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151092
CAS číslo: 68-19-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR