Tisk…

PF-04708671

Dodavatel: Apollo Scientific

Synonyms: PF-4708671, 2-((4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl)methyl)-6-(trifluoromethyl)-1H-benzo[d]imidazole

BISN0176-5MG
APOSBISN0176-5MGEA 744 CZK
APOSBISN0176-5MG
PF-04708671
PF-04708671

MAPK/ERK Signalling Pathway

Vzorec: C₁₉H₂₁F₃N₆
MW: 390.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18086922
CAS číslo: 1255517-76-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR