Tisk…

Y-27632 dihydrochloride

Dodavatel: Apollo Scientific

BISN0135-10MG BISN0135-100MG
APOSBISN0135-10MGEA 3310 CZK
APOSBISN0135-10MG APOSBISN0135-100MG
Y-27632 dihydrochloride
Y-27632 dihydrochloride

Y-27632 dihydrochloride is a selective ROCK1 p160ROCK inhibitor with Ki of 140 nM exhibits 200-fold selectivity over other kinases including PKC cAMP-dependent protein kinase MLCK and PAK.

Vzorec: C₁₄H₂₁N₃O·2HCl
Storage Temperature: Refrigerator
MDL Number: MFCD03490488
CAS číslo: 129830-38-2 | 146986-50-7 (free base)

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR