Tisk…

Bedaquiline

Dodavatel: Apollo Scientific

Synonyms: (1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol

BISN0131-5MG BISN0131-10MG
APOSBISN0131-5MGEA 3940 CZK
APOSBISN0131-5MG APOSBISN0131-10MG
Bedaquiline
Bedaquiline

Bedaquiline, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.

Vzorec: C₃₂H₃₁BrN₂O₂
MW: 555.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628872
CAS číslo: 843663-66-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR