Tisk…

Sulphosuccinimidyl 4-(4-azidophenyl)butyrate

Dodavatel: Apollo Scientific

BIPA115-100MG
APOSBIPA115-100MGEA 2690 CZK
APOSBIPA115-100MG
Sulphosuccinimidyl 4-(4-azidophenyl)butyrate
Sulphosuccinimidyl 4-(4-azidophenyl)butyrate

Photoreactive -Primary amine reactive Water soluble

Vzorec: C₁₄H₁₄N₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00153601
CAS číslo: 223572-55-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR