Tisk…

N-(tert-Butoxycarbonyl)-L-phenylalanine 98%

Dodavatel: Apollo Scientific
Varování

BIP0718-500G
APOSBIP0718-500GEA 2650 CZK
APOSBIP0718-500G
N-(tert-Butoxycarbonyl)-L-phenylalanine 98%
N-(tert-Butoxycarbonyl)-L-phenylalanine
Vzorec: C₁₄H₁₉NO₄
Bod tání: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002663
CAS číslo: 13734-34-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR