Tisk…

Penicillin G sodium salt

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

BIP0142-100G BIP0142-250G BIP0142-25G
APOSBIP0142-100GEA 1660 CZK
APOSBIP0142-100G APOSBIP0142-250G APOSBIP0142-25G
Penicillin G sodium salt
Penicillin G sodium salt

Penicillin G is an inhibitor of bacterial cell wall synthesis.Active against gram-positive and some gram-negative bacteria. b-lactamase sensitive.


  • Potency: 1670 mg/mg

Vzorec: C₁₆H₁₇N₂NaO₄S
MW: 356.38
Bod tání: 255
Storage Temperature: Refrigerator
MDL Number: MFCD00069666
CAS číslo: 69-57-8
EINECS: 200-710-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR