Tisk…

Neosolaniol (from Fusarium sp.)

Dodavatel: Apollo Scientific
Nebezpečí

BIN1002-1MG BIN1002-5MG
APOSBIN1002-1MGEA 5790 CZK
APOSBIN1002-1MG APOSBIN1002-5MG
Neosolaniol (from Fusarium sp.)
Neosolaniol
Vzorec: C₁₉H₂₆O₈
Bod tání: 176…178 °C
Storage Temperature: Freezer
MDL Number: MFCD16661260
CAS číslo: 36519-25-2
UN: 2811
ADR: 6.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR