Tisk…

Nile blue chloride (Basic blue 12)

Dodavatel: Apollo Scientific
Varování

BIN0464-10G BIN0464-25G BIN0464-100G
APOSBIN0464-10GEA 1240 CZK
APOSBIN0464-10G APOSBIN0464-25G APOSBIN0464-100G
Nile blue chloride (Basic blue 12)
Nile blue chloride (Basic blue 12)

Fluorecence reference standard.

Vzorec: C₂₀H₂₀ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD00011936
CAS číslo: 2381-85-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR