Tisk…

N-Octyl-β-D-glucopyranoside 99, ultrapure

Dodavatel: Apollo Scientific
Varování

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

BIMB1075-10G BIMB1075-1G BIMB1075-5G
APOSBIMB1075-10GEA 0 CZK
APOSBIMB1075-10G APOSBIMB1075-1G APOSBIMB1075-5G
N-Octyl-β-D-glucopyranoside 99, ultrapure
N-Octyl-β-D-glucopyranoside

Non-ionic detergent used for membrane protein solubisation. An advantage of OG is its low molecular weight which allows easy removal by dialysis.

Vzorec: C₁₄H₂₈O₆
MW: 292.37
Bod tání: 103…104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063288
CAS číslo: 29836-26-8
EINECS: 249-887-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR