Tisk…

Guanidiniumthiokyanát ≥99.0%

Dodavatel: Apollo Scientific
Nebezpečí

BIMB1010-1KG BIMB1010-500G BIMB1010-5KG
APOSBIMB1010-1KGEA 2900 CZK
APOSBIMB1010-1KG APOSBIMB1010-500G APOSBIMB1010-5KG
Guanidiniumthiokyanát ≥99.0%
Guanidiniumthiokyanát

Protein denaturant.


  • Toxic-LD50 of <500mg/Kg

Vzorec: CH₅N₃·CHNS
Bod tání: 117…122 °C
Hustota: 1,29 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013027
CAS číslo: 593-84-0
EINECS: 209-812-1
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR