Tisk…

Methyl-ɑ-D-mannopyranoside

Dodavatel: Apollo Scientific
Varování

BIM1008-100G BIM1008-25G BIM1008-500G
APOSBIM1008-100GEA 2480 CZK
APOSBIM1008-100G APOSBIM1008-25G APOSBIM1008-500G
Methyl-ɑ-D-mannopyranoside
Methyl-alpha-D-mannopyranoside
Vzorec: C₇H₁₄O₆
Bod tání: 186…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS číslo: 617-04-9
EINECS: 210-502-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR