Tisk…

L-Methionin ≥99%

Dodavatel: Apollo Scientific

BIM0715-100G BIM0715-1KG BIM0715-500G
APOSBIM0715-100GEA 622 CZK
APOSBIM0715-100G APOSBIM0715-1KG APOSBIM0715-500G
L-Methionin ≥99%
L-Methionin

Ph.Eur, Pyrogen free

Vzorec: C₅H₁₁NO₂S
Bod tání: 284 °C
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS číslo: 63-68-3
EINECS: 200-562-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR