Tisk…

Cefoxitin sodium

Dodavatel: Apollo Scientific
Varování

BIM0108-1G BIM0108-250MG BIM0108-5G
APOSBIM0108-1GEA 1870 CZK
APOSBIM0108-1G APOSBIM0108-250MG APOSBIM0108-5G
Cefoxitin sodium
Cefoxitin sodium
Vzorec: C₁₆H₁₆N₃NaO₇S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00079042
CAS číslo: 33564-30-6
EINECS: 251-574-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR