Tisk…

Mupirocin lithium

Dodavatel: Apollo Scientific

Synonyms: Lithium 9-(((E)-4-((2S,3R,4R,5S)-3,4-dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoate

BIL9900-100MG BIL9900-500MG BIL9900-1G
APOSBIL9900-100MGEA 1870 CZK
APOSBIL9900-100MG APOSBIL9900-500MG APOSBIL9900-1G
Mupirocin lithium
Mupirocin lithium
Vzorec: C₂₆H₄₃LiO₉
Storage Temperature: Ambient
MDL Number: MFCD11977847
CAS číslo: 73346-79-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR