Tisk…

Resveratrol

Dodavatel: Apollo Scientific
Varování

BIK9013-25G
APOSBIK9013-25GEA 662 CZK
APOSBIK9013-25G
Resveratrol
Resveratrol

20% or 99%

Vzorec: C₁₄H₁₂O₃
Bod tání: 261…267 °C
Storage Temperature: Freezer
MDL Number: MFCD00133799
CAS číslo: 501-36-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR