Tisk…

Elacridar

Dodavatel: Apollo Scientific
Varování

BIE3025-25MG BIE3025-50MG BIE3025-10MG
APOSBIE3025-25MGEA 3940 CZK
APOSBIE3025-25MG APOSBIE3025-50MG APOSBIE3025-10MG
Elacridar
Elacridar
Vzorec: C₃₄H₃₃N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00912604
CAS číslo: 143664-11-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR