Tisk…

Piperlongumine

Dodavatel: Apollo Scientific
Varování

BIDF1035-250MG BIDF1035-50MG BIDF1035-100MG
APOSBIDF1035-250MGEA 4960 CZK
APOSBIDF1035-250MG APOSBIDF1035-50MG APOSBIDF1035-100MG
Piperlongumine
Piperlongumine

Natural product.

Vzorec: C₁₇H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00075706
CAS číslo: 20069-09-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR