Tisk…

Dimethylsulfoxid, Cell Culture grade

Dodavatel: Apollo Scientific
Varování

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

BID1200-4X250M
APOSBID1200-4X250MEA 9490 CZK
APOSBID1200-4X250M
Dimethylsulfoxid, Cell Culture grade
Dimethylsulfoxid
Vzorec: (CH₃)₂SO
MW: 78.14
Bod varu: 189 °C
Bod tání: 18 °C
Hustota: 1.1 g/cm³ (25 °C)
Bod vzplanutí: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS číslo: 67-68-5
EINECS: 200-604-3
Merck Index: 13,03285

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR