Tisk…

Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4361-1G BICL4361-2G BICL4361-500MG
APOSBICL4361-1GEA 21890 CZK
APOSBICL4361-1G APOSBICL4361-2G APOSBICL4361-500MG
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside
Vzorec: C₂₇H₃₈O₁₈
Storage Temperature: Ambient
MDL Number: MFCD28397808
CAS číslo: 13223-83-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR