Tisk…

Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4283-250MG BICL4283-500MG
APOSBICL4283-250MGEA 17920 CZK
APOSBICL4283-250MG APOSBICL4283-500MG
Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside ≥98%
Methyl 4,6-dichloro-4,6-dideoxy-α-D-galactopyranoside
Vzorec: C₇H₁₂Cl₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08457713
CAS číslo: 4990-82-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR