Tisk…

Propargyl α-D-mannopyranoside ≥98%

Dodavatel: Apollo Scientific
Varování

BICL4262-100MG BICL4262-250MG BICL4262-500MG
APOSBICL4262-100MGEA 13960 CZK
APOSBICL4262-100MG APOSBICL4262-250MG APOSBICL4262-500MG
Propargyl α-D-mannopyranoside ≥98%
Propargyl α-D-mannopyranoside
Vzorec: C₉H₁₄O₆
Storage Temperature: Freezer
MDL Number: MFCD19980990
CAS číslo: 854262-01-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR