Tisk…

3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%

Dodavatel: Apollo Scientific

Synonyms: 3,4,6-Tri-o-benzyl-1,2-o-(1-methoxyethylidene)-beta-D-mannopyranose, (3aS,5R,6R,7S,7aS)-6,7-Bis(benzyloxy)-5-((benzyloxy)methyl)-2-methoxy-2-methyltetrahydro-3aH-[1,3]dioxolo[4,5-b]pyran, 3,4,6-Tri-O-benzyl-β-D-mannopyranose 1,2-(methyl orthoacetate)

BICL4218-1G
APOSBICL4218-1GEA 39350 CZK
APOSBICL4218-1G
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose
Vzorec: C₃₀H₃₄O₇
MW: 506.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270088
CAS číslo: 16697-49-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR