Tisk…

Methyl 4,6-O-benzylidene-β-D-galactopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4170-1G BICL4170-5G BICL4170-2G
APOSBICL4170-1GEA 6020 CZK
APOSBICL4170-1G APOSBICL4170-5G APOSBICL4170-2G
Methyl 4,6-O-benzylidene-β-D-galactopyranoside ≥98%
Methyl 4,6-O-benzylidene-β-D-galactopyranoside
Storage Temperature: Ambient
CAS číslo: 6988-39-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR