Tisk…

4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%

Dodavatel: Apollo Scientific

BICL4144-5G
APOSBICL4144-5GEA 2070 CZK
APOSBICL4144-5G
4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%
4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside
Vzorec: C₂₁H₂₆O₉S
Storage Temperature: Ambient
MDL Number: MFCD13182351
CAS číslo: 28244-94-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR