Tisk…

4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4143-1G BICL4143-5G BICL4143-2G
APOSBICL4143-1GEA 6780 CZK
APOSBICL4143-1G APOSBICL4143-5G APOSBICL4143-2G
4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 905716-02-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR