Tisk…

Benzoic acid-acyl-β-D-glucuronide ≥98%

Dodavatel: Apollo Scientific

BICL4036-1MG BICL4036-5MG BICL4036-2MG
APOSBICL4036-1MGEA 11180 CZK
APOSBICL4036-1MG APOSBICL4036-5MG APOSBICL4036-2MG
Benzoic acid-acyl-β-D-glucuronide ≥98%
Benzoic acid-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 19237-53-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR