Tisk…

Coomassie Brilliant Blue G-250

Dodavatel: Apollo Scientific
Varování

BIC3480-100G BIC3480-25G
APOSBIC3480-100GEA 3940 CZK
APOSBIC3480-100G APOSBIC3480-25G
Coomassie Brilliant Blue G-250
Coomassie Brilliant Blue G-250

C.I. 42655

Vzorec: C₄₇H₄₈N₃NaO₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS číslo: 6104-58-1
EINECS: 228-058-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR