Tisk…

Cyclopiazonic acid

Dodavatel: Apollo Scientific
Nebezpečí

BIC1012-5MG BIC1012-10MG BIC1012-25MG
APOSBIC1012-5MGEA 3110 CZK
APOSBIC1012-5MG APOSBIC1012-10MG APOSBIC1012-25MG
Cyclopiazonic acid
Cyclopiazonic acid
Vzorec: C₂₀H₂₀N₂O₃
MW: 336.391 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00167445
CAS číslo: 18172-33-3
UN: 2811
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR