Tisk…

N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine ≥98%

Dodavatel: Apollo Scientific

Synonyms: 1-(9H-Fluoren-9-yl)-3,19-dioxo-2,8,11,14-tetraoxa-4,18-diazadocosan-22-oic acid

BIBP1077-500MG BIBP1077-1G BIBP1077-5G
APOSBIBP1077-500MGEA 2280 CZK
APOSBIBP1077-500MG APOSBIBP1077-1G APOSBIBP1077-5G
N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine ≥98%
N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine
Vzorec: C₂₉H₃₈N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD05663769
CAS číslo: 172089-14-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR